Learn More
ATCC K562-r; Chronic myeloid leukemia ; Human (Homo sapiens)
Catalog No. 502385104
Encompass
K56S-r is a cell line with hematopoietic morphology that was isolated in 1999 from the bone marrow of a 53-year-old female with chronic myeloid leukemia at blast crisis. The paired K562-s and K562-r clones are useful cell line models of sensitivity and resistance to a tyrosine kinase inhibitor used in the treatment of chronic myeloid leukemia. It has applications for immune system disorder research.
Catalog No. 50-238-5104 Supplier ATCC Supplier No. CRL3344
K56S-r is a cell line with hematopoietic morphology that was isolated in 1999 from the bone marrow of a 53-year-old female with chronic myeloid leukemia at blast crisis. The paired K562-s and K562-r clones are useful cell line models of sensitivity and resistance to a tyrosine kinase inhibitor used in the treatment of chronic myeloid leukemia. It has applications for immune system disorder research.