Learn More
INTACT GENOMICS INC K599 Agrobacterium Chemically Competent Cells 18x50µl
Supplier:  INTACT GENOMICS INC 108718
Encompass
Intact Genomics (ig) K599 Agrobacterium Chemically Competent Cells are made from a specific strain of Agrobacterium rhizogenes, Agrobacterium rhizogenes (str R) pRi2659 (agropine type). K599 was originally isolated from cucumber exhibiting hairy root disease symptoms and has been widely used for hairy root transformation(1).Agrobacterium rhizogenes is a soil-borne gram-negative bacterium that can infect most dicotyledons, a few monocotyledons and some gymnosperms. K599 Chemically Competent Agrobacterium are optimized for the highest transformation efficiencies and are useful for transgenic operations of corn, soybean (wild soybean), cotton, peanut, dandelion, cowpea and other plants. K599 Agrobacterium rhizogenes strain contains pRi2659 agrobacterium-type Ri plasmid and displays Spectinomycin resistance.Product Includes and Storage: K599 Agrobacterium Chemically Competent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-82978
 Restricted Item