Learn More
INTACT GENOMICS INC K599 Agrobacterium ElectroCompetent Cells 6x50µl
Supplier:  INTACT GENOMICS INC 128712
Encompass
Intact Genomics (ig) K599 Agrobacterium ElectroCompetent Cells are made from a specific strain of Agrobacterium rhizogenes, Agrobacterium rhizogenes (str R) pRi2659 (agropine type). K599 was originally isolated from cucumber exhibiting hairy root disease symptoms and has been widely used for hairy root transformation (1).Agrobacterium rhizogenes is a soil-borne gram-negative bacterium that can infect most dicotyledons, a few monocotyledons, and some gymnosperms. IG’s K599 Agrobacterium ElectroCompetent Cells is optimized for the highest transformation efficiencies and is useful for transgenic operations of corn, soybean, cotton, peanut, dandelion, cowpea, and other plants. K599 Agrobacterium rhizogenes strain contains pRi2659 agrobacterium-type Ri plasmid and displays Spectinomycin resistance.Product Includes and Storage:K599 Agrobacterium ElectroCompetent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82869