Learn More
Bioss K63-linkage specific ubiquitin
SDP
Catalog No. 5000065213
Encompass_Preferred
K63-linkage specific ubiquitin Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65213 Supplier Bioss Supplier No. BSM63006R
K63-linkage specific ubiquitin Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol