Learn More
Grainger Steiner Welding Jacket, Flame Retardant Cotton, Mesh Back, 30 in., Large, 1 ea
Catalog No. 502758189
Encompass_Preferred
This 30 in. black welding jacket is designed for professional protection against flames, sparks, and metal spatter. Constructed from flame-retardant 100% cotton with a ventilated FR polyester mesh back, this garment offers a balance of safety and comfort for demanding work environments.
  • Flame-retardant 100% cotton construction
  • Ventilated mesh back for increased breathability
  • Includes 2 pockets and a dedicated soapstone pocket
  • Secure snap closure
  • Passes ASTM D6413-10 flame testing
  • Size: L
Catalog No. 50-275-8189 Supplier Grainger Supplier No. 154A33
This 30 in. black welding jacket is designed for professional protection against flames, sparks, and metal spatter. Constructed from flame-retardant 100% cotton with a ventilated FR polyester mesh back, this garment offers a balance of safety and comfort for demanding work environments.
  • Flame-retardant 100% cotton construction
  • Ventilated mesh back for increased breathability
  • Includes 2 pockets and a dedicated soapstone pocket
  • Secure snap closure
  • Passes ASTM D6413-10 flame testing
  • Size: L