Learn More
Grainger Basco Transport Drum, Closed Head, Polyethylene, 15 Gal., 1 Ea.
Catalog No. 502784201
Encompass_Preferred
This 15 gal. closed head transport drum is constructed from seamless, high-molecular-weight, high-density polyethylene, making it suitable for indoor and outdoor storage of liquids, food, chemicals, and hardware. The drum features a non-removable cover with two bung holes, a molded swing handle for portability, and a durable, molded-in blue color that resists denting, rusting, and fading.
  • BPA free, prime food grade material
  • FDA and USDA compliant
  • UN rated for shipping hazardous materials under DOT 49 CFR
  • Seamless one-piece mold construction
  • Equipped with a 2 in. NPS fitting and a 2 in. buttress fitting with a 3/4 in. NPT reducer
  • Load capacity of 237 lbs.
Catalog No. 50-278-4201 Supplier Grainger Supplier No. 19H053
This 15 gal. closed head transport drum is constructed from seamless, high-molecular-weight, high-density polyethylene, making it suitable for indoor and outdoor storage of liquids, food, chemicals, and hardware. The drum features a non-removable cover with two bung holes, a molded swing handle for portability, and a durable, molded-in blue color that resists denting, rusting, and fading.
  • BPA free, prime food grade material
  • FDA and USDA compliant
  • UN rated for shipping hazardous materials under DOT 49 CFR
  • Seamless one-piece mold construction
  • Equipped with a 2 in. NPS fitting and a 2 in. buttress fitting with a 3/4 in. NPT reducer
  • Load capacity of 237 lbs.