Learn More
Grainger BASCO Closed Head Plastic Drum, HDPE, 55 Gal., Black, 1 ea.
Catalog No. 502783928
Encompass_Preferred
This 55 gal. closed head transport drum is constructed from durable, unlined HDPE. Designed for industrial use, this drum is suitable for handling chemicals, coatings, and solvents.
  • Manufactured from polyethylene
  • Black body and cover color
  • Features two bung holes for standard closures
  • UN/DOT rated for hazardous service
  • Load capacity of 870 lb.
  • Measures 59.18 cm (23.3 in.) in diameter and 88.39 cm (34.8 in.) in height
  • Material thickness of 0.14 in.
Catalog No. 50-278-3928 Supplier Grainger Supplier No. 19H058
This 55 gal. closed head transport drum is constructed from durable, unlined HDPE. Designed for industrial use, this drum is suitable for handling chemicals, coatings, and solvents.
  • Manufactured from polyethylene
  • Black body and cover color
  • Features two bung holes for standard closures
  • UN/DOT rated for hazardous service
  • Load capacity of 870 lb.
  • Measures 59.18 cm (23.3 in.) in diameter and 88.39 cm (34.8 in.) in height
  • Material thickness of 0.14 in.