Learn More
TARGETMOL CHEMICALS INC K777 5MG
SDP
Catalog No. 502581214
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K777 is a potent orally active and irreversible inhibitor of cysteine protease. K777 is a potent CYP3A4 inhibitor (IC50= 60 nM). K777 is also a selective CCR4 antagonist featuring the potent chemotaxis inhibition. K777 irreversibly inhibits Cruzain which is the major cysteine protease of Trypansoma cruzi and cathepsins B and L. K777 targets cathepsin-mediated cell entry and wxibits a broad-spectrum antiviral activity. K777 inhibits EBOV and SARS-CoV pseudovirus entry with IC50 values of 0.87 nM and 0.68 nM respectively. purity: 99%
Catalog No. 50-258-1214 Supplier TARGETMOL CHEMICALS INC Supplier No. T156415MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K777 is a potent orally active and irreversible inhibitor of cysteine protease. K777 is a potent CYP3A4 inhibitor (IC50= 60 nM). K777 is also a selective CCR4 antagonist featuring the potent chemotaxis inhibition. K777 irreversibly inhibits Cruzain which is the major cysteine protease of Trypansoma cruzi and cathepsins B and L. K777 targets cathepsin-mediated cell entry and wxibits a broad-spectrum antiviral activity. K777 inhibits EBOV and SARS-CoV pseudovirus entry with IC50 values of 0.87 nM and 0.68 nM respectively. purity: 99%