Learn More
Eurofins Discoverx [Lys8,9]-Neurotensin (8-13), 5 mg. (92-1059)
Supplier: Eurofins Discoverx 921059
[Lys8,9]-Neurotensin (8-13) is the smallest active fragment of Neurotensin, a 13 amino acid neuropeptide of the CNS that is implicated in luteinizing hormone (LH) and dopamine regulation, and prolactin release. Effects of Neurotensin activity include vasodilation, analgesia, hypothermia, and increased locomotion. This [Lys8,9]-analog of neurotensin enhances amiloride-sensitive Na+-dependent transmembrane H+ flux. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.