Learn More
Medchemexpress LLC K858 (Racemic) | 72926-24-0 | 99.8% | 277.35 | 5 MG
SDP
Catalog No. 5000393433
Encompass_Preferred
K858 (Racemic) is an ATP-uncompetitive inhibitor of kinesin Eg5, with an IC50 of 1.3 μM. It specifically induces mitotic arrest and growth inhibition by activating the Mad2-mediated spindle checkpoint, leading to mitotic cell death in cancer cells without affecting normal cells. It also shows antitumor activity in ovarian and colon cancer xenograft models in vivo.
  • ATP-uncompetitive inhibitor of kinesin Eg5
  • Induces mitotic arrest and growth inhibition
  • Activates the Mad2-mediated spindle checkpoint
  • Induces mitotic cell death in cancer cells at 5 μM
  • Shows antitumor activity in A2780 ovarian and HCT116 colon cancer xenograft models
  • Displays no neurotoxic side effects in mice
Catalog No. 50-003-93433 Supplier Medchemexpress LLC Supplier No. 5MGHY19966
K858 (Racemic) is an ATP-uncompetitive inhibitor of kinesin Eg5, with an IC50 of 1.3 μM. It specifically induces mitotic arrest and growth inhibition by activating the Mad2-mediated spindle checkpoint, leading to mitotic cell death in cancer cells without affecting normal cells. It also shows antitumor activity in ovarian and colon cancer xenograft models in vivo.
  • ATP-uncompetitive inhibitor of kinesin Eg5
  • Induces mitotic arrest and growth inhibition
  • Activates the Mad2-mediated spindle checkpoint
  • Induces mitotic cell death in cancer cells at 5 μM
  • Shows antitumor activity in A2780 ovarian and HCT116 colon cancer xenograft models
  • Displays no neurotoxic side effects in mice