Learn More
Accuflow Systems Saint-Gobain Sani-Tech Concentric Reducer, PVDF, 1.5 in x 3/4 in
Catalog No. NC3440541
Encompass_Preferred
The Saint-Gobain Sani-Tech concentric reducer KAD150-075 is a sanitary PVDF reducer that provides a concentric reduction from 1.5 in to 3/4 in for bioprocess and sterile fluid handling systems.
  • Sanitary concentric reduction from 1.5 in to 3/4 in.
  • Manufactured from PVDF for chemical resistance and dimensional stability.
  • Concentric configuration for inline flow transition and minimal dead volume.
  • Dimension A (small end): 0.750 in; dimension B: 1.606 in; dimension C: 3.000 in.
  • Available in polypropylene or PVDF materials across the Sani-Tech concentric reducer family.
  • Designed for sanitary and bioprocessing applications where smooth internal surfaces reduce entrapment.
Catalog No. NC3440541 Supplier Accuflow Systems Supplier No. KAD150075
The Saint-Gobain Sani-Tech concentric reducer KAD150-075 is a sanitary PVDF reducer that provides a concentric reduction from 1.5 in to 3/4 in for bioprocess and sterile fluid handling systems.
  • Sanitary concentric reduction from 1.5 in to 3/4 in.
  • Manufactured from PVDF for chemical resistance and dimensional stability.
  • Concentric configuration for inline flow transition and minimal dead volume.
  • Dimension A (small end): 0.750 in; dimension B: 1.606 in; dimension C: 3.000 in.
  • Available in polypropylene or PVDF materials across the Sani-Tech concentric reducer family.
  • Designed for sanitary and bioprocessing applications where smooth internal surfaces reduce entrapment.