Learn More
MSC SPI Carbide Contact Point, 0.040 in. Diameter, 7/16 in. Length, 1 ea.
Catalog No. 5000516691
Encompass
This carbide contact point is designed for precision measurement applications in industrial settings, providing reliable performance for inspection and metrology tasks.
  • Constructed from durable carbide for extended wear resistance
  • Compatible with standard indicator systems
  • Engineered for consistent and accurate contact measurements
Catalog No. 50-005-16691 Supplier MSC Supplier No. 01746270
This carbide contact point is designed for precision measurement applications in industrial settings, providing reliable performance for inspection and metrology tasks.
  • Constructed from durable carbide for extended wear resistance
  • Compatible with standard indicator systems
  • Engineered for consistent and accurate contact measurements