Learn More
Cayman Chemical KaempferIde 10mg
SDP
Catalog No. 502988781
Encompass_Preferred
A flavonoid with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 97.58 μg/ml); cytotoxic to HT-1080 and Colon 26-L5 cells (EC50s = 2.91 and 5.95 μg/ml, respectively); decreases lipid accumulation in 3T3-L1 preadipocytes at 50 μM; attenuates decreases in LVSP and increases in LVEDP and reduces infarct size in a rat model of myocardial ischemia-reperfusion injury induced by coronary artery ligation at 0.3 and 1 mg/kg
Catalog No. 50-298-8781 Supplier Cayman Chemical Supplier No. 2967310 MG
A flavonoid with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 97.58 μg/ml); cytotoxic to HT-1080 and Colon 26-L5 cells (EC50s = 2.91 and 5.95 μg/ml, respectively); decreases lipid accumulation in 3T3-L1 preadipocytes at 50 μM; attenuates decreases in LVSP and increases in LVEDP and reduces infarct size in a rat model of myocardial ischemia-reperfusion injury induced by coronary artery ligation at 0.3 and 1 mg/kg