Learn More
Medchemexpress LLC Kaempferide | 491-54-3 | C16H12O6 | 1 ML
SDP
Catalog No. 5000376690
Encompass_Preferred
Kaempferide is an orally active flavonol isolated from Hippophae rhamnoides L.. It exhibits a broad spectrum of biological activities, including anticancer, anti-inflammatory, antioxidant, antidiabetic, antiobesity, antihypertensive, and neuroprotective effects. The compound induces apoptosis and promotes osteogenesis, making it valuable for osteoporosis research.
  • Orally active flavonol
  • Exhibiting anticancer, anti-inflammatory, antioxidant, antidiabetic, antiobesity, antihypertensive, and neuroprotective activities
  • Induces apoptosis and promotes osteogenesis
  • Reduces lipid accumulation and oxidative stress
  • Inhibits melanogenesis
  • Ameliorates oxidative stress and inflammation in obese models
  • Alleviates myocardial ischemia/reperfusion injury
Catalog No. 50-003-76690 Supplier Medchemexpress LLC Supplier No. HY1544910MM/1ML
Kaempferide is an orally active flavonol isolated from Hippophae rhamnoides L.. It exhibits a broad spectrum of biological activities, including anticancer, anti-inflammatory, antioxidant, antidiabetic, antiobesity, antihypertensive, and neuroprotective effects. The compound induces apoptosis and promotes osteogenesis, making it valuable for osteoporosis research.
  • Orally active flavonol
  • Exhibiting anticancer, anti-inflammatory, antioxidant, antidiabetic, antiobesity, antihypertensive, and neuroprotective activities
  • Induces apoptosis and promotes osteogenesis
  • Reduces lipid accumulation and oxidative stress
  • Inhibits melanogenesis
  • Ameliorates oxidative stress and inflammation in obese models
  • Alleviates myocardial ischemia/reperfusion injury