Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaempferide, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014307 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4307 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4307 Supplier MilliporeSigma Supelco Supplier No. 6954510MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kaempferide is a naturally occurring, O-methylated flavonol found in Kaempferia galanga. It exhibits a variety of pharmacological effects such as anti-inflammatory, anti-viral, anti-oxidant, anti-bacterial and antitumor properties.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 491-54-3
Physical Form Neat
Linear Formula C16H12O6
MDL Number MFCD00016771
Synonym 4′-Methoxy-3,5,7-trihydroxyflavone
Type Analytical Standard
Quantity 10 mg
Formula Weight 300.26
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.