Learn More
TARGETMOL CHEMICALS INC KAEMPFERITRIN 5MG
SDP
Catalog No. 502576952
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kaempferitrin (Lespenephryl) has antidepressant-like effect related to the serotonergic system principally 5-HT1A. Kaempferitrin exerts cytotoxic and antitumor effects against HeLa cells. Kaempferitrin stimulates glucose-metabolizing enzymes promotes glucose homeostasis. Kaempferitrin exerts immunostimulatory effects on immune responses mediated by splenocytes macrophages PBMC and NK cells. Kaempferitrin induces cytotoxic effects to include cell cycle arrest in G1 phase and apoptosis via the intrinsic pathway in a caspase-dependent pathway. purity: 99%
Catalog No. 50-257-6952 Supplier TARGETMOL CHEMICALS INC Supplier No. T33865MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kaempferitrin (Lespenephryl) has antidepressant-like effect related to the serotonergic system principally 5-HT1A. Kaempferitrin exerts cytotoxic and antitumor effects against HeLa cells. Kaempferitrin stimulates glucose-metabolizing enzymes promotes glucose homeostasis. Kaempferitrin exerts immunostimulatory effects on immune responses mediated by splenocytes macrophages PBMC and NK cells. Kaempferitrin induces cytotoxic effects to include cell cycle arrest in G1 phase and apoptosis via the intrinsic pathway in a caspase-dependent pathway. purity: 99%