Learn More
Indofine Chemical Kaempferol, 99%, 100 Mg
SDP
Catalog No. 50908550
Encompass_Preferred
KAEMPFEROL 100 MG, Flavone, 3,5,7,4'-Tetrahydroxyflavone; Robigenin, C15H10O6, MW: 286.24, 0.99, 520-18-3, MFCD00016938
Catalog No. 50-908-550 Supplier Indofine Chemical Supplier No. K102 100 MG
KAEMPFEROL 100 MG, Flavone, 3,5,7,4'-Tetrahydroxyflavone; Robigenin, C15H10O6, MW: 286.24, 0.99, 520-18-3, MFCD00016938