Learn More
Apexbio Technology LLC Kaempferol(Synonyms: Kempferol, Kaempherol, Kaempferide, Robigenin, Rhamnolutein, Populnetin), 10mM (in 1mL DMSO), CAS: 520-18-3.
SDP
Catalog No. 5000061142
Encompass_Preferred
Kaempferol (CAS 520-18-3) is a naturally derived flavonoid isolated from sources including Gingko biloba and red wine It functions biologically by activating the mitochondrial calcium uniporter (EC50 7 M) Additionally kaempferol promotes apoptosis through the caspase-9 pathway by suppressing polo-like kinase 1 (PLK1) expression thereby inhibiting cancer cell proliferation in diverse cell lines It also demonstrates antioxidant properties and reduces osteoclast-mediated bone resorption in vitro These characteristics make kaempferol valuable for researching mitochondrial physiology cancer biology and bone metabolism
Catalog No. 50-000-61142 Supplier Apexbio Technology LLC Supplier No. N17195.1
Kaempferol (CAS 520-18-3) is a naturally derived flavonoid isolated from sources including Gingko biloba and red wine It functions biologically by activating the mitochondrial calcium uniporter (EC50 7 M) Additionally kaempferol promotes apoptosis through the caspase-9 pathway by suppressing polo-like kinase 1 (PLK1) expression thereby inhibiting cancer cell proliferation in diverse cell lines It also demonstrates antioxidant properties and reduces osteoclast-mediated bone resorption in vitro These characteristics make kaempferol valuable for researching mitochondrial physiology cancer biology and bone metabolism