Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kaempferol 3,7,4'-trimethyl ether | 15486-34-7 | 5 MG
SDP

Catalog No. 5000419992
Encompass_Preferred

Kaempferol 3,7,4'-trimethyl ether is a flavonol aglycone isolated from the leaves of *Siparuna gigantotepala*. It exhibits antioxidant activity and is intended for research use only.

  • 98.0% purity
  • Molecular weight: 328.32
  • Chemical formula: C18H16O6
  • Solid appearance
  • Light yellow to yellow color
  • Classified as a flavonoid, flavonol, phenol, and monophenol
  • Isolated from plants
  • Demonstrates cytotoxicity against human A549, HT-29, and HeLa cells

Catalog No. 50-004-19992 Supplier Medchemexpress LLC Supplier No. HYN34345MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kaempferol 3,7,4'-trimethyl ether is a flavonol aglycone isolated from the leaves of *Siparuna gigantotepala*. It exhibits antioxidant activity and is intended for research use only.

  • 98.0% purity
  • Molecular weight: 328.32
  • Chemical formula: C18H16O6
  • Solid appearance
  • Light yellow to yellow color
  • Classified as a flavonoid, flavonol, phenol, and monophenol
  • Isolated from plants
  • Demonstrates cytotoxicity against human A549, HT-29, and HeLa cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.