Learn More
Medchemexpress LLC Kaempferol 3,7,4'-trimethyl ether | 15486-34-7 | 98.0% | 328.32 g/mol | C18H16O6 | 10 MG
SDP
Catalog No. 5000289611
Encompass_Preferred
Kaempferol 3,7,4'-trimethyl ether is a naturally occurring flavonol aglycone isolated from the leaves of Siparuna gigantotepala. It is supplied as a research-grade reference compound and is reported to exhibit antioxidant activity. The compound has formula C18H16O6, molecular weight 328.32 g/mol, CAS 15486-34-7, and is provided with 98.0% purity.
  • Naturally derived flavonol aglycone
  • Exhibits antioxidant activity
  • High purity: 98.0%
  • Molecular formula C18H16O6; molecular weight 328.32 g/mol
  • Supplied as a solid reference for research applications
  • Available in small pack sizes suitable for compound screening and analytical work
Catalog No. 50-002-89611 Supplier Medchemexpress LLC Supplier No. HYN343410MG
Kaempferol 3,7,4'-trimethyl ether is a naturally occurring flavonol aglycone isolated from the leaves of Siparuna gigantotepala. It is supplied as a research-grade reference compound and is reported to exhibit antioxidant activity. The compound has formula C18H16O6, molecular weight 328.32 g/mol, CAS 15486-34-7, and is provided with 98.0% purity.
  • Naturally derived flavonol aglycone
  • Exhibits antioxidant activity
  • High purity: 98.0%
  • Molecular formula C18H16O6; molecular weight 328.32 g/mol
  • Supplied as a solid reference for research applications
  • Available in small pack sizes suitable for compound screening and analytical work