Learn More
TARGETMOL CHEMICALS INC Kaempferol 3-O-beta-D-glucuron
SDP
Catalog No. 502875826
Encompass_Preferred
Also available in 1 mL, 1 mg, 50 mg and bulk. Please contact Fisher for quotes. Kaempferol 3-O-beta-D-glucuronide (Kaempferol-3-beta-O-glucuronide) exhibits antioxidant activity. Flavonoid glucuronides can be deconjugated by microsomal alpha²-glucuronidase from various human cells. Purity 99.07%
Catalog No. 50-287-5826 Supplier TARGETMOL CHEMICALS INC Supplier No. TN18185MG
Also available in 1 mL, 1 mg, 50 mg and bulk. Please contact Fisher for quotes. Kaempferol 3-O-beta-D-glucuronide (Kaempferol-3-beta-O-glucuronide) exhibits antioxidant activity. Flavonoid glucuronides can be deconjugated by microsomal alpha²-glucuronidase from various human cells. Purity 99.07%