Learn More
Cayman Chemical Kaempferol 3-O-galactoside, Purity: ≥98%, CAS: 23627-87-4, 5 mg
SDP
Catalog No. 502948292
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits sPLA2 (IC50 = 17.6 µM); decreases protein levels of Akt and induces apoptosis in NCI H460 NSCLC cells at 12.5, 25, and 50 µM; scavenges DPPH radicals and superoxide anions in cell-free assays at 200 µg/ml; reduces TPA-induced ear edema and carrageenan-induced paw edema in mice at 150 mg/kg
Catalog No. 50-294-8292 Supplier Cayman Chemical Supplier No. 337475 MG
A flavonoid with diverse biological activities; inhibits sPLA2 (IC50 = 17.6 µM); decreases protein levels of Akt and induces apoptosis in NCI H460 NSCLC cells at 12.5, 25, and 50 µM; scavenges DPPH radicals and superoxide anions in cell-free assays at 200 µg/ml; reduces TPA-induced ear edema and carrageenan-induced paw edema in mice at 150 mg/kg