Learn More
Medchemexpress LLC HY-N0208 5mg Medchemexpress, Kaempferol-3-O-glucorhamnoside CAS:40437-72-7 Purity:>98%
SDP
Catalog No. 501970235
Encompass_Preferred
Medchemexpress, HY-N0208 5mg Kaempferol-3-O-glucorhamnoside CAS:40437-72-7 Kaempferol-3-O-glucorhamnoside, a flavonoid derived from plant Thesium chinense Turcz, inhibits inflammatory responses via MAPK and NF-κB pathways in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0235 Supplier Medchemexpress LLC Supplier No. HYN02085MG
Medchemexpress, HY-N0208 5mg Kaempferol-3-O-glucorhamnoside CAS:40437-72-7 Kaempferol-3-O-glucorhamnoside, a flavonoid derived from plant Thesium chinense Turcz, inhibits inflammatory responses via MAPK and NF-κB pathways in vitro and in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.