Learn More
Selleck Chemical LLC Kaempferol-3-O-glucorhamnoside S5468-1mg
SDP
Catalog No. 503111537
Encompass_Preferred
Kaempferol 3-neohesperidoside (Kaempferol 3-neohesperidoside) isolated from the herbs of Delphinium grandiflorum L has insulin-like properties in terms of glucose lowering
Catalog No. 50-311-1537 Supplier Selleck Chemical LLC Supplier No. S54681MG
Kaempferol 3-neohesperidoside (Kaempferol 3-neohesperidoside) isolated from the herbs of Delphinium grandiflorum L has insulin-like properties in terms of glucose lowering