Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kaempferol 3-O-rutinoside 7-O-glucoside | 34336-18-0 | 1 MG
SDP

Catalog No. 5000419374
Encompass_Preferred

Kaempferol 3-O-rutinoside 7-O-glucoside is a flavonoid glycoside found in red tomato. This product is for research use only.

  • Purity: 99.67%
  • Molecular weight: 756.66
  • Formula: C₃₃H₄₀O₂₀
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Store at 4°C, sealed, away from moisture
  • Store in solvent: -80°C for 6 months, or -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-19374 Supplier Medchemexpress LLC Supplier No. HYN81651MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kaempferol 3-O-rutinoside 7-O-glucoside is a flavonoid glycoside found in red tomato. This product is for research use only.

  • Purity: 99.67%
  • Molecular weight: 756.66
  • Formula: C₃₃H₄₀O₂₀
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Store at 4°C, sealed, away from moisture
  • Store in solvent: -80°C for 6 months, or -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.