Learn More
Medchemexpress LLC Kaempferol | 520-18-3 | MFCD00004543 | 99.9% | C15H10O6 | 200 MG
SDP
Catalog No. 5000356131
Encompass_Preferred
Kaempferol is a naturally occurring flavonoid found in various edible plants. This compound functions as an estrogen receptor alpha inhibitor in breast cancer cells and promotes apoptosis in glioblastoma and lung cancer cells through MEK-MAPK activation. It is recognized as a human endogenous metabolite.
  • Inhibits estrogen receptor alpha expression in breast cancer cells.
  • Induces apoptosis in glioblastoma and lung cancer cells.
  • Demonstrates anti-inflammatory activity.
  • Exhibits antioxidant activity in human HepG2 cells.
  • Shows antiproliferative activity against certain human cancer cell lines.
  • Displays antiallergic activity.
  • Demonstrates antiviral activity against influenza A virus.
  • Inhibits melanogenesis.
  • Inhibits procoagulant activity.
  • Induces osteoclast apoptosis.
  • Exhibits antiaggregatory activity in human platelets.
Catalog No. 50-003-56131 Supplier Medchemexpress LLC Supplier No. HY14590200MG
Kaempferol is a naturally occurring flavonoid found in various edible plants. This compound functions as an estrogen receptor alpha inhibitor in breast cancer cells and promotes apoptosis in glioblastoma and lung cancer cells through MEK-MAPK activation. It is recognized as a human endogenous metabolite.
  • Inhibits estrogen receptor alpha expression in breast cancer cells.
  • Induces apoptosis in glioblastoma and lung cancer cells.
  • Demonstrates anti-inflammatory activity.
  • Exhibits antioxidant activity in human HepG2 cells.
  • Shows antiproliferative activity against certain human cancer cell lines.
  • Displays antiallergic activity.
  • Demonstrates antiviral activity against influenza A virus.
  • Inhibits melanogenesis.
  • Inhibits procoagulant activity.
  • Induces osteoclast apoptosis.
  • Exhibits antiaggregatory activity in human platelets.