Learn More
Medchemexpress LLC Kaempferol-7,4'-dimethyl ether | 15486-33-6 | MFCD00017441 | 314.29 | 1 MG
SDP
Catalog No. 5000415422
Encompass_Preferred
Kaempferol-7,4'-dimethyl ether is an inhibitor of PTP1B (Protein Tyrosine Phosphatase 1B) and also inhibits the production of nitric oxide (NO). It exhibits an inhibition rate of 46.1% at 100 μM and an IC50 value of 16.92 μM for PTP1B activity. This product is for research use only and is not sold to patients.
  • PTP1B inhibitor
  • Inhibits nitric oxide production
  • Exhibits 46.1% inhibition at 100 μM
  • Has an IC50 value of 16.92 μM for PTP1B activity
Catalog No. 50-004-15422 Supplier Medchemexpress LLC Supplier No. HYN17891MG
Kaempferol-7,4'-dimethyl ether is an inhibitor of PTP1B (Protein Tyrosine Phosphatase 1B) and also inhibits the production of nitric oxide (NO). It exhibits an inhibition rate of 46.1% at 100 μM and an IC50 value of 16.92 μM for PTP1B activity. This product is for research use only and is not sold to patients.
  • PTP1B inhibitor
  • Inhibits nitric oxide production
  • Exhibits 46.1% inhibition at 100 μM
  • Has an IC50 value of 16.92 μM for PTP1B activity