Learn More
TARGETMOL CHEMICALS INC Kaempferol-7 4 -dimethyl ether
SDP
Catalog No. 502877073
Encompass_Preferred
Also available in 1 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Kaempferol-7,4'-dimethyl ether, isolated from the rhizomes of Zingiber phillipseae, exhibits antioxidant activity against S. aureus and E. coli with MICs of 200 and 225 ug/ml, respectively. Purity 98.54%
Catalog No. 50-287-7073 Supplier TARGETMOL CHEMICALS INC Supplier No. TN28982MG
Also available in 1 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Kaempferol-7,4'-dimethyl ether, isolated from the rhizomes of Zingiber phillipseae, exhibits antioxidant activity against S. aureus and E. coli with MICs of 200 and 225 ug/ml, respectively. Purity 98.54%