Learn More
TARGETMOL CHEMICALS INC KAEMPFEROL-7-O-BETA-D-GL 5MG
SDP
Catalog No. 502579973
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kaempferol-7-O-(beta)-D-glucopyranoside (Kaempferol-7-O-D-glucopyranoside) a novel supercoolant could be used to sub-zero non-freezing rat heart preservation. purity: 99%
Catalog No. 50-257-9973 Supplier TARGETMOL CHEMICALS INC Supplier No. T40735MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kaempferol-7-O-(beta)-D-glucopyranoside (Kaempferol-7-O-D-glucopyranoside) a novel supercoolant could be used to sub-zero non-freezing rat heart preservation. purity: 99%