Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kaempferol-7-O-β-D-glucopyranoside | 16290-07-6 | 448.38 | 1 MG
SDP

Catalog No. 5000389111
Encompass_Preferred

Kaempferol-7-O-β-D-glucopyranoside is a flavonoid glycoside isolated from *Malus pumila Mill.* flowers and also found in the root bark of *Cudrania tricuspidata*. It is intended for research use only. This compound exhibits antioxidant, anti-inflammatory, and procoagulant activities, including the scavenging of DPPH free radicals and inhibition of Cu2+-induced lipid peroxidation of low-density lipoprotein (LDL).

  • Exhibits antioxidative activity
  • Exhibits anti-inflammatory activity
  • Exhibits procoagulant activity
  • Scavenges DPPH free radicals
  • Inhibits Cu2+-induced lipid peroxidation of LDL
  • Purity of 99.79%
  • Appearance is solid, light yellow to yellow

Catalog No. 50-003-89111 Supplier Medchemexpress LLC Supplier No. HYN06271MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kaempferol-7-O-β-D-glucopyranoside is a flavonoid glycoside isolated from *Malus pumila Mill.* flowers and also found in the root bark of *Cudrania tricuspidata*. It is intended for research use only. This compound exhibits antioxidant, anti-inflammatory, and procoagulant activities, including the scavenging of DPPH free radicals and inhibition of Cu2+-induced lipid peroxidation of low-density lipoprotein (LDL).

  • Exhibits antioxidative activity
  • Exhibits anti-inflammatory activity
  • Exhibits procoagulant activity
  • Scavenges DPPH free radicals
  • Inhibits Cu2+-induced lipid peroxidation of LDL
  • Purity of 99.79%
  • Appearance is solid, light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.