Learn More
Medchemexpress LLC Kaempferol-7-O-β-D-glucopyranoside | 16290-07-6 | 448.38 | 5 MG
SDP
Catalog No. 5000389060
Encompass_Preferred
Kaempferol-7-O-β-D-glucopyranoside is a flavonoid glycoside found in the root bark of *Cudrania tricuspidata*. It has demonstrated antioxidant activity by inhibiting Cu2+-induced lipid peroxidation of low-density lipoprotein and by scavenging DPPH free radicals.
  • Inhibits lipid peroxidation.
  • Possesses antioxidant properties.
  • Scavenges free radicals.
Catalog No. 50-003-89060 Supplier Medchemexpress LLC Supplier No. HYN06275MG
Kaempferol-7-O-β-D-glucopyranoside is a flavonoid glycoside found in the root bark of *Cudrania tricuspidata*. It has demonstrated antioxidant activity by inhibiting Cu2+-induced lipid peroxidation of low-density lipoprotein and by scavenging DPPH free radicals.
  • Inhibits lipid peroxidation.
  • Possesses antioxidant properties.
  • Scavenges free radicals.