Learn More
Medchemexpress LLC HY-N3431 5mg Medchemexpress, Kaempferol-7-O-rhamnoside CAS:20196-89-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN34315MG
Encompass_Preferred
Medchemexpress, HY-N3431 5mg Kaempferol-7-O-rhamnoside CAS:20196-89-8 Kaempferol-7-O-rhamnoside, isolated from Chimonanthus nitens Oliv. Leaves, is a potent α-glucosidase activity inhibitor. Kaempferol-7-O-rhamnoside has the potential for diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2913