Learn More
Abcam Kaempferol, Phytoestrogen, 50MG
Catalog No. 502707655
Encompass_Preferred
MW 286.24 Da, Purity >98%. Cell-permeable phytoestrogen and dietary flavonoid. Proapoptotic and protects against amyloid beta peptide neurotoxicity in vivo. Has many biological actions including: inhibition of topoisomerase-1, MAO COX, estrogenic effects and activation of the mitochondrial Ca²+ uniporter.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7655 Supplier Abcam Supplier No. AB12035750MG
MW 286.24 Da, Purity >98%. Cell-permeable phytoestrogen and dietary flavonoid. Proapoptotic and protects against amyloid beta peptide neurotoxicity in vivo. Has many biological actions including: inhibition of topoisomerase-1, MAO COX, estrogenic effects and activation of the mitochondrial Ca²+ uniporter.
The product is subject to the following: Abcam Restricted Use Statement