Learn More
Cayman Chemical KaempfRol 3nohSpRIdosID 10mg
SDP
Catalog No. 502950483
Encompass_Preferred
A flavonoid with antioxidant and antidiabetic activities; scavenges DPPH radicals (IC50 = 79.6 UG/ml); increases glucose uptake in isolated rat soleus muscle at 1 and 100 nM; increases glycogen synthesis in isolated rat soleus muscle at 1 µM; reduces serum glucose levels in a rat model of alloxan-induced diabetes at 100 mg/kg
Catalog No. 50-295-0483 Supplier Cayman Chemical Supplier No. 3453310 MG
A flavonoid with antioxidant and antidiabetic activities; scavenges DPPH radicals (IC50 = 79.6 UG/ml); increases glucose uptake in isolated rat soleus muscle at 1 and 100 nM; increases glycogen synthesis in isolated rat soleus muscle at 1 µM; reduces serum glucose levels in a rat model of alloxan-induced diabetes at 100 mg/kg