Learn More
Cayman Chemical Kaempferol 7-neohesperidoside, Purity: ≥95%, CAS: 17353-03-6, 10 mg
SDP
Catalog No. 502911288
Encompass_Preferred
A flavonoid glycoside with antioxidant and anticancer activities; scavenges DPPH radicals in a cell-free assay (IC50 = 7.4 µM); cytotoxic to A549, HepG2, and HeLa cells (IC50s = 0.53, 0.2, and 0.051 µM, respectively)
Catalog No. 50-291-1288 Supplier Cayman Chemical Supplier No. 3483110 MG
A flavonoid glycoside with antioxidant and anticancer activities; scavenges DPPH radicals in a cell-free assay (IC50 = 7.4 µM); cytotoxic to A549, HepG2, and HeLa cells (IC50s = 0.53, 0.2, and 0.051 µM, respectively)