Learn More
Enzo Life Sciences Kahweol (10mg). CAS: 6894-43-5
SDP
Supplier:  Enzo Life Sciences ALX350223M010
Encompass_Preferred
Induces glutathione S-transferase. Has anti-cancer, chemoprotective, anti-inflammatory and anti-atherosclerotic properties. Purity: ≥97% (UPLC). Solubility: Soluble in ethyl acetate, acetone or DMSO; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8785