Learn More
FUJIFILM Biosciences Ham's F12K (Kaighn's Nutrient Mixture F12) - Liquid
Catalog No. NC1404526
Encompass
Ham's F12K (Kaighn's Nutrient Mixture F12) was originally developed for use in cloning Chinese hamster ovary andLung cells. This medium is useful with serum supplements for a variety of cell culture applications, including the growth of myeloma and hybridoma cells.
Catalog No. NC1404526 Supplier FUJIFILM Biosciences Supplier No. 90771L
Ham's F12K (Kaighn's Nutrient Mixture F12) was originally developed for use in cloning Chinese hamster ovary andLung cells. This medium is useful with serum supplements for a variety of cell culture applications, including the growth of myeloma and hybridoma cells.