Learn More
Enzo Life Sciences Kainic acid (25mg). CAS: 487-79-6
SDP
Supplier:  Enzo Life Sciences BMLEA1230025
Encompass_Preferred
Selective agonist at kainate receptors. Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0531