Learn More
Medchemexpress LLC Kainic acid hydrate | 58002-62-3 | 25 MG
SDP
Catalog No. 5000421556
Encompass_Preferred
Kainic acid hydrate is a potent excitotoxic agent and an agonist for a subtype of ionotropic glutamate receptor. It is recognized for its ability to induce seizures and can be utilized to create epilepsy models. This compound can be administered systemically, into the hippocampus, or amygdala, showing reproducible results across various species. The systemic Kainic acid model closely mimics human temporal lobe epilepsy.
  • Potent excitotoxic agent
  • Agonist for a subtype of ionotropic glutamate receptor
  • Induces seizures
  • Utilized to create epilepsy models
  • Reproducible in various species
  • Systemic model mimics human temporal lobe epilepsy
Catalog No. 50-004-21556 Supplier Medchemexpress LLC Supplier No. 25MGHYN2309A
Add to Cart
Add to Cart
Kainic acid hydrate is a potent excitotoxic agent and an agonist for a subtype of ionotropic glutamate receptor. It is recognized for its ability to induce seizures and can be utilized to create epilepsy models. This compound can be administered systemically, into the hippocampus, or amygdala, showing reproducible results across various species. The systemic Kainic acid model closely mimics human temporal lobe epilepsy.
  • Potent excitotoxic agent
  • Agonist for a subtype of ionotropic glutamate receptor
  • Induces seizures
  • Utilized to create epilepsy models
  • Reproducible in various species
  • Systemic model mimics human temporal lobe epilepsy