Learn More
BIOGEMS INTERNATIONAL INC Kainic Acid Monohydrate 50mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 580623250MG
Encompass
Kainic acid is a potent neuroexcitatory amino acid that acts as a neurotoxin in high doses. It was originally isolated from seaweed and is used in the study of the physiological effect of excitotoxicity and assess the neuroprotective capabilities of potential therapeutics.
Catalog No. 50-153-0190