Learn More
Abcam Kainic acid, prototypic kainate receptor agonist, 1MG
Catalog No. 502584697
Encompass_Preferred
MW 213.23 Da, Purity >99%. Prototypic agonist at the kainate class of ionotropic glutamate receptors. Potent excitant and neurotoxin, used to model epilepsy and neurodegenerative states.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4697 Supplier Abcam Supplier No. AB1201001MG
MW 213.23 Da, Purity >99%. Prototypic agonist at the kainate class of ionotropic glutamate receptors. Potent excitant and neurotoxin, used to model epilepsy and neurodegenerative states.
The product is subject to the following: Abcam Restricted Use Statement