Learn More
Waters Corp Application Kit, Kairos Amino Acid Analysis, Low Throughput, Manual
Catalog No. 502393000 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This application kit is designed for accurate and precise amino acid analysis in plasma and urine samples, intended for research use only. It provides detailed step-by-step instructions for manual use.
  • Designed for accurate and precise amino acid analysis in plasma and urine samples.
  • Intended for research use only.
  • Features verified AccQ·Tag™ derivatization chemistry.
  • Compatible with UPLC I-Class PLUS System, Xevo TQ-S micro, and QDa Detector systems.
  • Offers low throughput (100+).
  • Derivatized samples are stable for one week at room temperature in a sealed vial, allowing for retesting.
Catalog No. 50-239-3000 Supplier Waters Corp Supplier No. 176004375
Add to Cart
Add to Cart
Request a Quote
This application kit is designed for accurate and precise amino acid analysis in plasma and urine samples, intended for research use only. It provides detailed step-by-step instructions for manual use.
  • Designed for accurate and precise amino acid analysis in plasma and urine samples.
  • Intended for research use only.
  • Features verified AccQ·Tag™ derivatization chemistry.
  • Compatible with UPLC I-Class PLUS System, Xevo TQ-S micro, and QDa Detector systems.
  • Offers low throughput (100+).
  • Derivatized samples are stable for one week at room temperature in a sealed vial, allowing for retesting.