Learn More
Waters Corp Application Kit, Amino Acid, 500+, Automation, UPLC
Catalog No. 502393026 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for automated sample preparation workflows for amino acid quantification. It is a first-time order kit (500+) for high-throughput, LC-MS-based amino acid analysis in plasma and urine samples, providing accurate and precise results, quantifying over 40 amino acids with rapid 9-minute assays. This kit is for research use only.
  • Automated sample preparation workflows
  • Accurate and precise amino acid analysis
  • Quantifies over 40 amino acids
  • Compatible with automated liquid handling platforms
  • Uses AccQ·Tag derivatization chemistry
  • Utilizes UPLC and mass spectrometry detection
  • Includes calibrators, quality controls, and internal standards
  • Suitable for plasma and urine samples
  • Offers rapid 9-minute assays
Catalog No. 50-239-3026 Supplier Waters Corp Supplier No. 176004377
Add to Cart
Add to Cart
Request a Quote
This kit is designed for automated sample preparation workflows for amino acid quantification. It is a first-time order kit (500+) for high-throughput, LC-MS-based amino acid analysis in plasma and urine samples, providing accurate and precise results, quantifying over 40 amino acids with rapid 9-minute assays. This kit is for research use only.
  • Automated sample preparation workflows
  • Accurate and precise amino acid analysis
  • Quantifies over 40 amino acids
  • Compatible with automated liquid handling platforms
  • Uses AccQ·Tag derivatization chemistry
  • Utilizes UPLC and mass spectrometry detection
  • Includes calibrators, quality controls, and internal standards
  • Suitable for plasma and urine samples
  • Offers rapid 9-minute assays