Learn More
Waters Corp Kairos Amino Acid Kit, High Throughput, Automation, Plasma and Urine Analysis
Catalog No. 502390949 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Kairos Amino Acid Kit – High Throughput (500+) Automation is a research-use-only kit designed for accurate and precise amino acid analysis in plasma and urine samples. Optimized for automation with liquid handling platforms, offering streamlined high-volume sample preparation and analysis.
  • High throughput for analyzing over 500 samples.
  • Automation ready for hands-free sample preparation.
  • Accurate and precise amino acid analysis.
  • Scalable reagents for automated platforms.
  • Streamlined workflow with step-by-step protocol.
  • Robust derivatization using AccQ·Tag chemistry.
  • Stable derivatized samples for one week.
  • Internal standards for reliable quantitation.
  • Optimized LC and MS methods provided.
Catalog No. 50-239-0949 Supplier Waters Corp Supplier No. 176004378
Add to Cart
Add to Cart
Request a Quote
The Waters Kairos Amino Acid Kit – High Throughput (500+) Automation is a research-use-only kit designed for accurate and precise amino acid analysis in plasma and urine samples. Optimized for automation with liquid handling platforms, offering streamlined high-volume sample preparation and analysis.
  • High throughput for analyzing over 500 samples.
  • Automation ready for hands-free sample preparation.
  • Accurate and precise amino acid analysis.
  • Scalable reagents for automated platforms.
  • Streamlined workflow with step-by-step protocol.
  • Robust derivatization using AccQ·Tag chemistry.
  • Stable derivatized samples for one week.
  • Internal standards for reliable quantitation.
  • Optimized LC and MS methods provided.