Learn More
Waters Corp Application Kit, Kairos Amino Acid, 500+, Manual First Time Order
Catalog No. 502392902 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Kairos Amino Acid 500+ Manual First Time Order Kit is for accurate and precise amino acid analysis in plasma and urine samples. This kit is for research use only, not for diagnostic procedures. Methods have been verified using AccQ·Tag™ derivatization chemistry and UPLC-MS systems such as ACQUITY™ UPLC™ I-Class PLUS System with Xevo™ TQ-S micro, and ACQUITY™ QDa™ Detector.
  • Designed for accurate and precise amino acid analysis in plasma and urine samples.
  • Features a workflow including protein precipitation, derivatization, LC-MS analysis, and data analysis.
  • Achieve a total preparation time for 96 samples of less than 2 hours.
  • Provides a rapid run time of 9 minutes.
  • Includes calibrator, quality control, internal standard sets, a UPLC C18 column, a derivatization kit, reconstitution reagents, formic acid, and LCMS vials.
Catalog No. 50-239-2902 Supplier Waters Corp Supplier No. 176004379
Add to Cart
Add to Cart
Request a Quote
The Kairos Amino Acid 500+ Manual First Time Order Kit is for accurate and precise amino acid analysis in plasma and urine samples. This kit is for research use only, not for diagnostic procedures. Methods have been verified using AccQ·Tag™ derivatization chemistry and UPLC-MS systems such as ACQUITY™ UPLC™ I-Class PLUS System with Xevo™ TQ-S micro, and ACQUITY™ QDa™ Detector.
  • Designed for accurate and precise amino acid analysis in plasma and urine samples.
  • Features a workflow including protein precipitation, derivatization, LC-MS analysis, and data analysis.
  • Achieve a total preparation time for 96 samples of less than 2 hours.
  • Provides a rapid run time of 9 minutes.
  • Includes calibrator, quality control, internal standard sets, a UPLC C18 column, a derivatization kit, reconstitution reagents, formic acid, and LCMS vials.