Learn More
Waters Corp Sample Preparation Workflow, Amino Acid, UPLC, 500+ Manual Re-order
Catalog No. 502391135 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Kairos Amino Acid 500+ Manual Re-order Kit is a sample preparation workflow designed for accurate quantitation of over 40 physiological amino acids in plasma and urine samples. It utilizes UPLC separation and mass spectrometry-based detection, offering a rapid 9-minute assay.
  • Rapid 9-minute assay.
  • Utilizes multiple stable labeled internal standards and mass spectrometry-based detection.
  • Simplifies retesting; samples can be re-injected into LC-MS.
  • Quantifies over 40 physiological amino acids in plasma and urine samples.
  • Applicable for research use only.
  • Verified for use with multiple automated liquid handling platforms.
  • Includes a CORTECS UPLC C18 column.
  • Derivatized samples are stable for one week at room temperature.
  • Total preparation time for 96 samples is less than 2 hours.
Catalog No. 50-239-1135 Supplier Waters Corp Supplier No. 176004380
Add to Cart
Add to Cart
Request a Quote
The Waters Kairos Amino Acid 500+ Manual Re-order Kit is a sample preparation workflow designed for accurate quantitation of over 40 physiological amino acids in plasma and urine samples. It utilizes UPLC separation and mass spectrometry-based detection, offering a rapid 9-minute assay.
  • Rapid 9-minute assay.
  • Utilizes multiple stable labeled internal standards and mass spectrometry-based detection.
  • Simplifies retesting; samples can be re-injected into LC-MS.
  • Quantifies over 40 physiological amino acids in plasma and urine samples.
  • Applicable for research use only.
  • Verified for use with multiple automated liquid handling platforms.
  • Includes a CORTECS UPLC C18 column.
  • Derivatized samples are stable for one week at room temperature.
  • Total preparation time for 96 samples is less than 2 hours.