Learn More
Waters Corp Calibrator Set, High-throughput (100+), Amino Acid Analysis, Mass Accuracy
Catalog No. 502390773 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This calibrator set is designed for high-throughput (100+) physiological amino acid analysis. It improves the detector’s mass-to-charge ratio, ensuring mass accuracy for amino acid analysis applications. Acting as a tuning compound, it provides spectral lines for the necessary operational mass range and helps prevent mass axis drift in QToF mass analyzers.
  • Configured for high-throughput (100+) physiological amino acid analysis.
  • Ensures mass accuracy by improving detector’s mass-to-charge ratio.
  • Undergoes stringent QC inspection for proper composition.
  • Functions as a tuning compound with spectral lines for operational mass range.
  • Contains all calibrators for the amino acid analysis workflow.
  • Helps prevent mass axis drift, essential for QToF mass analyzer users.
Catalog No. 50-239-0773 Supplier Waters Corp Supplier No. 186009193
This calibrator set is designed for high-throughput (100+) physiological amino acid analysis. It improves the detector’s mass-to-charge ratio, ensuring mass accuracy for amino acid analysis applications. Acting as a tuning compound, it provides spectral lines for the necessary operational mass range and helps prevent mass axis drift in QToF mass analyzers.
  • Configured for high-throughput (100+) physiological amino acid analysis.
  • Ensures mass accuracy by improving detector’s mass-to-charge ratio.
  • Undergoes stringent QC inspection for proper composition.
  • Functions as a tuning compound with spectral lines for operational mass range.
  • Contains all calibrators for the amino acid analysis workflow.
  • Helps prevent mass axis drift, essential for QToF mass analyzer users.