Learn More
Waters Corp Kairos Amino Acid Internal Standard Set, Isotopically Labeled, Physiological Analysis
Catalog No. 502392033 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This internal standard set is designed for accurate physiological amino acid analysis in plasma and urine samples. It helps accommodate analyte loss, corrects matrix effects, and serves as an internal control for quantitation within the Kairos Amino Acid Analysis workflow.
  • Isotopically labeled for accurate quantitation.
  • Contains sulfosalicylic acid for protein removal.
  • Compatible with advanced UPLC systems and mass spectrometry.
  • For research use only, not diagnostic procedures.
  • Optimizes amino acid analysis workflow and corrects matrix effects.
  • Includes LC-MS standards mirroring chemical species.
Catalog No. 50-239-2033 Supplier Waters Corp Supplier No. 186009051
This internal standard set is designed for accurate physiological amino acid analysis in plasma and urine samples. It helps accommodate analyte loss, corrects matrix effects, and serves as an internal control for quantitation within the Kairos Amino Acid Analysis workflow.
  • Isotopically labeled for accurate quantitation.
  • Contains sulfosalicylic acid for protein removal.
  • Compatible with advanced UPLC systems and mass spectrometry.
  • For research use only, not diagnostic procedures.
  • Optimizes amino acid analysis workflow and corrects matrix effects.
  • Includes LC-MS standards mirroring chemical species.