Learn More
Waters Corp Quality Control Set, Amino Acid, Physiological, 100+, LC-MS
Catalog No. 502392872 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Kairos Amino Acid Quality Control Set (100+) is designed for high-throughput physiological amino acid analysis using LC-MS systems. It provides standards to verify the performance of both liquid chromatography and mass spectrometry systems for amino acid detection in biological samples such as plasma and urine. Each standard is developed under strict quality control criteria to ensure reliable and consistent results.
  • Designed for high-throughput (100+) physiological amino acid analysis.
  • Provides comprehensive performance verification for LC-MS systems.
  • Developed under strict quality control criteria for reliable results.
  • Intended for research use only, not for diagnostic procedures.
  • Validated for use with plasma and urine samples.
  • Storage conditions: -10°C to -25°C.
Catalog No. 50-239-2872 Supplier Waters Corp Supplier No. 186009194
The Kairos Amino Acid Quality Control Set (100+) is designed for high-throughput physiological amino acid analysis using LC-MS systems. It provides standards to verify the performance of both liquid chromatography and mass spectrometry systems for amino acid detection in biological samples such as plasma and urine. Each standard is developed under strict quality control criteria to ensure reliable and consistent results.
  • Designed for high-throughput (100+) physiological amino acid analysis.
  • Provides comprehensive performance verification for LC-MS systems.
  • Developed under strict quality control criteria for reliable results.
  • Intended for research use only, not for diagnostic procedures.
  • Validated for use with plasma and urine samples.
  • Storage conditions: -10°C to -25°C.