Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Kairos Amino Acid Reconstitution Reagents, UPLC, 2 pk

Catalog No. 502392522 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These are reconstitution reagents specifically designed for physiological amino acid analysis.

  • Sample preparation workflow type
  • UPLC separation mode
  • As required replacement schedule
  • Amino acid application
  • Application kit product type
  • 2 units per package
  • Contains hazardous materials and requires special handling, which may incur additional charges.

Catalog No. 50-239-2522 Supplier Waters Corp Supplier No. 186009103
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These are reconstitution reagents specifically designed for physiological amino acid analysis.

  • Sample preparation workflow type
  • UPLC separation mode
  • As required replacement schedule
  • Amino acid application
  • Application kit product type
  • 2 units per package
  • Contains hazardous materials and requires special handling, which may incur additional charges.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.